Tisk…

2,6-Dichloro-4-fluorobenzyl bromide 95%

Dodavatel: Apollo Scientific
Nebezpečí

PC502275-250MG PC502275-1G
APOSPC502275-250MGEA 5790 CZK
APOSPC502275-250MG APOSPC502275-1G
2,6-Dichloro-4-fluorobenzyl bromide 95%
2,6-Dichloro-4-fluorobenzyl bromide
Vzorec: C₇H₄BrCl₂F
MW: 257.92 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28346557
CAS číslo: 1803784-34-0
UN: 3261
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR