Tisk…

Bis(dibenzylideneacetone)palladium 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: Bis(dibenzylideneacetone)palladium(0)

IN2826-1G IN2826-5G
APOSIN2826-1GEA 1660 CZK
APOSIN2826-1G APOSIN2826-5G
Bis(dibenzylideneacetone)palladium 98%
Bis(dibenzylideneacetone)palladium
Vzorec: C₃₄H₂₈O₂Pd
MW: 575 g/mol
Bod tání: 150 °C
MDL Number: MFCD00051942
CAS číslo: 32005-36-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR