Tisk…

(E)-Ethyl 3-iodoacrylate 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: Ethyl (E)-3-iodoprop-2-enoate

OR907663-1G OR907663-5G OR907663-10G
APOSOR907663-1GEA 2400 CZK
APOSOR907663-1G APOSOR907663-5G APOSOR907663-10G
(E)-Ethyl 3-iodoacrylate 97%
(E)-Ethyl 3-iodoacrylate
Vzorec: C₅H₇IO₂
MW: 226,01 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11519164
CAS číslo: 31930-37-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR