Tisk…

4-Hexylphenylboronic acid 97%

Dodavatel: Apollo Scientific
Varování

OR7224-1G OR7224-5G OR7224-250MG
APOSOR7224-1GEA 834 CZK
APOSOR7224-1G APOSOR7224-5G APOSOR7224-250MG
4-Hexylphenylboronic acid 97%
4-Hexylphenylboronic acid
Vzorec: C₁₂H₁₉BO₂
MDL Number: MFCD04039029
CAS číslo: 105365-50-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR