Tisk…

2-Cyano-phenothiazine

Dodavatel: Apollo Scientific
Varování

OR53126-25G OR53126-100G
APOSOR53126-25GEA 1660 CZK
APOSOR53126-25G APOSOR53126-100G
2-Cyano-phenothiazine
2-Cyano-phenothiazine
Vzorec: C₁₃H₈N₂S
Storage Temperature: Refrigerator
MDL Number: MFCD00129784
CAS číslo: 38642-74-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR