Tisk…

Isochroman-1-ylmethanamine 95%

Dodavatel: Apollo Scientific
Nebezpečí

OR322556-100MG OR322556-250MG OR322556-50MG
APOSOR322556-100MGEA 3310 CZK
APOSOR322556-100MG APOSOR322556-250MG APOSOR322556-50MG
Isochroman-1-ylmethanamine 95%
Isochroman-1-ylmethanamine
Vzorec: C₁₀H₁₃NO
MDL Number: MFCD00457645
CAS číslo: 19158-90-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR