meso-2,3-Dimercaptosuccinic acid ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
(R,S)-2,3-Dimercaptobutanedioic acid, meso-2,3-Dimercaptobutanedioic acid, Succimer, (R,S)-2,3-Dimercaptosuccinic acid, (2R,3S)-rel-2,3-Dimercaptosuccinic acid
A17909.06
A17909.03
A17909.06EA
3510
CZK
A17909.06
A17909.03
meso-2,3-Dimercaptosuccinic acid ≥97%
meso-2,3-Dimercaptosuccinic acid
Purity: 97%
CAS No.: 304-55-2
Molecular Formula: C4H6O4S2
Formula Weight: 182.22
Melting Point: ca 196° dec.
Beilstein Reference: 1725150
Merck Reference: 14,8865
MDL No.: MFCD00064799
Vzorec:
C₄H₆O₄S₂ MW: 182,22 g/mol Bod tání: ∼ 196 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00064799 CAS číslo: 304-55-2 EINECS: 206-155-2 Merck Index: 14,8865 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...