2-Amino-4-fluoro-3-(trifluoromethyl)benzonitrile 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC49369-250MG
PC49369-1G
APOSPC49369-250MGEA
3940
CZK
APOSPC49369-250MG
APOSPC49369-1G
2-Amino-4-fluoro-3-(trifluoromethyl)benzonitrile 95%
2-Amino-4-fluoro-3-(trifluoromethyl)benzonitrile
Vzorec:
C₈H₄F₄N₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD24850042 CAS číslo: 1440535-22-7 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...