Tisk…

1-(Cyclopropyl)-5-(trifluoromethyl)-1H-pyrazole-3-carboxylic acid 95%

Dodavatel: Apollo Scientific
Varování

PC408444-250MG PC408444-1G PC408444-5G
APOSPC408444-250MGEA 6220 CZK
APOSPC408444-250MG APOSPC408444-1G APOSPC408444-5G
1-(Cyclopropyl)-5-(trifluoromethyl)-1H-pyrazole-3-carboxylic acid 95%
1-(Cyclopropyl)-5-(trifluoromethyl)-1H-pyrazole-3-carboxylic acid
Vzorec: C₈H₇F₃N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD32669838
CAS číslo: 2384171-56-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR