Tisk…

4-Ethylthiophenol

Dodavatel: Apollo Scientific
Nebezpečí

OR11377-1G OR11377-25G
APOSOR11377-1GEA 1580 CZK
APOSOR11377-1G APOSOR11377-25G
4-Ethylthiophenol
4-Ethylthiophenol
Vzorec: C₈H₁₀S
Bod varu: 119 - 120/30 mm °C
Hustota: 1,02
Storage Temperature: Refrigerator
MDL Number: MFCD00040229
CAS číslo: 4946-13-8
UN: 2810
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR