Methyl 2-amino-6-bromobenzoate 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR916421-250MG
OR916421-1G
OR916421-5G
APOSOR916421-250MGEA
1080
CZK
APOSOR916421-250MG
APOSOR916421-1G
APOSOR916421-5G
Methyl 2-amino-6-bromobenzoate 97%
Methyl 2-amino-6-bromobenzoate
Vzorec:
C₈H₈BrNO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD11977360 CAS číslo: 135484-78-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...