Tisk…

2-((2-Methylbut-3-yn-2-yl)oxy)tetrahydro-2H-pyran 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-(2-Methylbut-3-yn-2-yloxy)oxane

OR70332-1G OR70332-5G OR70332-25G
APOSOR70332-1GEA 622 CZK
APOSOR70332-1G APOSOR70332-5G APOSOR70332-25G
2-((2-Methylbut-3-yn-2-yl)oxy)tetrahydro-2H-pyran 97%
2-((2-Methylbut-3-yn-2-yl)oxy)tetrahydro-2H-pyran
Vzorec: C₁₀H₁₆O₂
MW: 168.23 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00090526
CAS číslo: 27943-46-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR