Tisk…

7-Bromo-4-cyanoquinoline 95%

Dodavatel: Apollo Scientific
Varování

OR70307-250MG OR70307-1G
APOSOR70307-250MGEA 6220 CZK
APOSOR70307-250MG APOSOR70307-1G
7-Bromo-4-cyanoquinoline 95%
7-Bromo-4-cyanoquinoline
Vzorec: C₁₀H₅BrN₂
Storage Temperature: Refrigerator
MDL Number: MFCD32862641
CAS číslo: 1537610-35-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR