Tisk…

2-Bromo-3-thiophenecarbonitrile 95%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2-Bromothiophene-3-carbonitrile

OR12257-1G OR12257-250MG OR12257-5G
APOSOR12257-1GEA 5230 CZK
APOSOR12257-1G APOSOR12257-250MG APOSOR12257-5G
2-Bromo-3-thiophenecarbonitrile 95%
2-Bromo-3-thiophenecarbonitrile
Vzorec: C₅H₂BrNS
MW: 188,05 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09064981
CAS číslo: 56182-43-5
UN: 3276
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR