Tisk…

Methyl-1H-benzotriazole (mixture) 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: Tolyltriazole, 6(or7)-Methyl-1H-benzo[d][1,2,3]triazole, Methyl-1H-benzotriazole (mixture)

OR931649-100G OR931649-500G
APOSOR931649-100GEA 827 CZK
APOSOR931649-100G APOSOR931649-500G
Methyl-1H-benzotriazole (mixture) 98%
Methyl-1H-benzotriazole (mixture)
Vzorec: C₇H₇N₃
MW: 133,15 g/mol
Bod tání: 87 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00167158
CAS číslo: 29385-43-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR