Tisk…

N4-Benzoylcytidine 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: N-Benzoylcytidine

OR926177-5G OR926177-25G
APOSOR926177-5GEA 827 CZK
APOSOR926177-5G APOSOR926177-25G
N4-Benzoylcytidine 97%
N4-Benzoylcytidine
Vzorec: C₁₆H₁₇N₃O₆
Storage Temperature: Refrigerator
MDL Number: MFCD00010572
CAS číslo: 13089-48-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR