Tisk…

3,4-Difluoro-2-methoxyphenol 97%

Dodavatel: Apollo Scientific
Nebezpečí

PC45180-500MG PC45180-1G PC45180-5G
APOSPC45180-500MGEA 4370 CZK
APOSPC45180-500MG APOSPC45180-1G APOSPC45180-5G
3,4-Difluoro-2-methoxyphenol 97%
3,4-Difluoro-2-methoxyphenol
Vzorec: C₇H₆F₂O₂
MDL Number: MFCD17015349
CAS číslo: 158626-90-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR