Tisk…

Bis(lauroyloxy)dioctyltin 96%

Dodavatel: Apollo Scientific
Varování

Synonyms: [Dodecanoyloxy(dioctyl)stannyl] dodecanoate

OR918257-1G OR918257-25G OR918257-100G
APOSOR918257-1GEA 622 CZK
APOSOR918257-1G APOSOR918257-25G APOSOR918257-100G
Bis(lauroyloxy)dioctyltin 96%
Bis(lauroyloxy)dioctyltin
Vzorec: C₄₀H₈₀O₄Sn
MW: 743,77 g/mol
MDL Number: MFCD00026557
CAS číslo: 3648-18-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR