Tisk…

4-Bromo-2-fluoro-5-(trifluoromethyl)benzyl alcohol 95%

Dodavatel: Apollo Scientific
Varování

PC305031-250MG PC305031-1G
APOSPC305031-250MGEA 6220 CZK
APOSPC305031-250MG APOSPC305031-1G
4-Bromo-2-fluoro-5-(trifluoromethyl)benzyl alcohol 95%
4-Bromo-2-fluoro-5-(trifluoromethyl)benzyl alcohol
Vzorec: C₈H₅BrF₄O
MW: 273.02 g/mol
MDL Number: MFCD33395092
CAS číslo: 2092603-19-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR