3-Ethoxy-N-methyl-2-nitroaniline
Dodavatel: Apollo Scientific
|
Varování
|
OR72155-1G
OR72155-5G
APOSOR72155-1GEA
12340
CZK
APOSOR72155-1G
APOSOR72155-5G
3-Ethoxy-N-methyl-2-nitroaniline
3-Ethoxy-N-methyl-2-nitroaniline
Vzorec:
C₉H₁₂N₂O₃ |
MDL Number:
MFCD23993724 CAS číslo: 1515219-57-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...