3,5-Dimethyl-2-nitroaniline 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR62164-250MG
OR62164-1G
OR62164-5G
APOSOR62164-250MGEA
4960
CZK
APOSOR62164-250MG
APOSOR62164-1G
APOSOR62164-5G
3,5-Dimethyl-2-nitroaniline 95%
3,5-Dimethyl-2-nitroaniline
Vzorec:
C₈H₁₀N₂O₂ MW: 166.18 g/mol |
MDL Number:
MFCD18824861 CAS číslo: 35490-74-5 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...