Tisk…

2-Bromo-3,6-difluorobenzyl bromide 95%

Dodavatel: Apollo Scientific
Nebezpečí

PC501813-250MG PC501813-1G PC501813-5G
APOSPC501813-250MGEA 5790 CZK
APOSPC501813-250MG APOSPC501813-1G APOSPC501813-5G
2-Bromo-3,6-difluorobenzyl bromide 95%
2-Bromo-3,6-difluorobenzyl bromide
Vzorec: C₇H₄Br₂F₂
MW: 285.91 g/mol
MDL Number: MFCD28061450
CAS číslo: 1805119-92-9
UN: 3261
ADR: 8 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR