2-Bromo-5-fluoro-1-iodo-3-(trifluoromethyl)benzene 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC501309-250MG
PC501309-1G
APOSPC501309-250MGEA
6390
CZK
APOSPC501309-250MG
APOSPC501309-1G
2-Bromo-5-fluoro-1-iodo-3-(trifluoromethyl)benzene 95%
2-Bromo-5-fluoro-1-iodo-3-(trifluoromethyl)benzene
Vzorec:
C₇H₂BrF₄I MW: 368.89 g/mol |
MDL Number:
MFCD28348581 CAS číslo: 1934934-44-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...