Tisk…

5-Bromo-2-chloro-4-fluorobenzonitrile 95%

Dodavatel: Apollo Scientific
Nebezpečí

PC47466-250MG PC47466-1G PC47466-5G
APOSPC47466-250MGEA 3540 CZK
APOSPC47466-250MG APOSPC47466-1G APOSPC47466-5G
5-Bromo-2-chloro-4-fluorobenzonitrile 95%
5-Bromo-2-chloro-4-fluorobenzonitrile
Vzorec: C₇H₂BrClFN
CAS číslo: 1892285-40-3
UN: 2811
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR