Tisk…

4-Bromo-2-fluoro-5-(trifluoromethyl)benzonitrile 95%

Dodavatel: Apollo Scientific
Nebezpečí

PC305029-250MG PC305029-1G PC305029-5G
APOSPC305029-250MGEA 4960 CZK
APOSPC305029-250MG APOSPC305029-1G APOSPC305029-5G
4-Bromo-2-fluoro-5-(trifluoromethyl)benzonitrile 95%
4-Bromo-2-fluoro-5-(trifluoromethyl)benzonitrile
Vzorec: C₈H₂BrF₄N
MW: 268.01 g/mol
MDL Number: MFCD32265190
CAS číslo: 2091422-93-2
UN: 2811
ADR: 6.1 I

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR