Tisk…

4-Bromo-5-fluoro-2-iodobenzyl bromide 95%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 1-Bromo-4-(bromomethyl)-2-fluoro-5-iodobenzene

PC501571-250MG PC501571-1G PC501571-5G
APOSPC501571-250MGEA 4960 CZK
APOSPC501571-250MG APOSPC501571-1G APOSPC501571-5G
4-Bromo-5-fluoro-2-iodobenzyl bromide 95%
4-Bromo-5-fluoro-2-iodobenzyl bromide
Vzorec: C₇H₄Br₂FI
MW: 393.82 g/mol
MDL Number: MFCD18910205
CAS číslo: 1379330-38-7
UN: 3265
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR