Tisk…

2-Amino-5-cyclopropylbenzoic acid 95%

Dodavatel: Apollo Scientific
Varování

OR70327-250MG OR70327-1G OR70327-5G
APOSOR70327-250MGEA 4960 CZK
APOSOR70327-250MG APOSOR70327-1G APOSOR70327-5G
2-Amino-5-cyclopropylbenzoic acid 95%
2-Amino-5-cyclopropylbenzoic acid
Vzorec: C₁₀H₁₁NO₂
MDL Number: MFCD06802840
CAS číslo: 68701-47-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR