2-Amino-3-cyclopropylbenzoic acid 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR70325-250MG
OR70325-1G
OR70325-5G
APOSOR70325-250MGEA
4960
CZK
APOSOR70325-250MG
APOSOR70325-1G
APOSOR70325-5G
2-Amino-3-cyclopropylbenzoic acid 95%
2-Amino-3-cyclopropylbenzoic acid
Vzorec:
C₁₀H₁₁NO₂ MW: 177,2 g/mol |
MDL Number:
MFCD32862614 CAS číslo: 2654087-87-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...