4-Bromo-3-cyclopropylbenzoic acid 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR70323-250MG
OR70323-1G
OR70323-5G
APOSOR70323-250MGEA
4960
CZK
APOSOR70323-250MG
APOSOR70323-1G
APOSOR70323-5G
4-Bromo-3-cyclopropylbenzoic acid 95%
4-Bromo-3-cyclopropylbenzoic acid
Vzorec:
C₁₀H₉BrO₂ |
MDL Number:
MFCD32862612 CAS číslo: 2384580-34-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...