2-Bromo-3-cyclopropylbenzoic acid 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR70316-250MG
OR70316-1G
OR70316-5G
APOSOR70316-250MGEA
4470
CZK
APOSOR70316-250MG
APOSOR70316-1G
APOSOR70316-5G
2-Bromo-3-cyclopropylbenzoic acid 95%
2-Bromo-3-cyclopropylbenzoic acid
Vzorec:
C₁₀H₉BrO₂ MW: 241,08 g/mol |
MDL Number:
MFCD32862618 CAS číslo: 2654087-88-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...