Tisk…

2-Bromo-5-cyclopropylbenzoic acid 99%

Dodavatel: Apollo Scientific
Varování

OR70315-250MG OR70315-1G
APOSOR70315-250MGEA 4930 CZK
APOSOR70315-250MG APOSOR70315-1G
2-Bromo-5-cyclopropylbenzoic acid 99%
2-Bromo-5-cyclopropylbenzoic acid
Vzorec: C₁₀H₉BrO₂
MW: 241,08 g/mol
MDL Number: MFCD29067871
CAS číslo: 1692662-86-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR