Tisk…

3-Bromo-5-chloro-2-iodobenzoic acid 95%

Dodavatel: Apollo Scientific
Varování

OR61183-250MG OR61183-1G OR61183-5G
APOSOR61183-250MGEA 4960 CZK
APOSOR61183-250MG APOSOR61183-1G APOSOR61183-5G
3-Bromo-5-chloro-2-iodobenzoic acid 95%
3-Bromo-5-chloro-2-iodobenzoic acid
Vzorec: C₇H₃BrClIO₂
MDL Number: MFCD07780679
CAS číslo: 213771-13-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR