4-Methoxy-2,3,5,6-tetrafluorophenylboronic acid
Dodavatel: Apollo Scientific
|
Varování
|
PC412512-5G
PC412512-10G
PC412512-25G
APOSPC412512-5GEA
16530
CZK
APOSPC412512-5G
APOSPC412512-10G
APOSPC412512-25G
4-Methoxy-2,3,5,6-tetrafluorophenylboronic acid
4-Methoxy-2,3,5,6-tetrafluorophenylboronic acid
Vzorec:
C₇H₅BF₄O₃ MW: 223,92 g/mol |
MDL Number:
MFCD07784386 CAS číslo: 871126-20-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...