6-Methyl-3,4-dihydro-2H-1,4-benzoxazine 98%
Dodavatel: Apollo Scientific
|
Varování
|
OR949546-100MG
OR949546-250MG
OR949546-1G
APOSOR949546-100MGEA
4100
CZK
APOSOR949546-100MG
APOSOR949546-250MG
APOSOR949546-1G
6-Methyl-3,4-dihydro-2H-1,4-benzoxazine 98%
6-Methyl-3,4-dihydro-2H-1,4-benzoxazine
Vzorec:
C₉H₁₁NO |
MDL Number:
MFCD01935457 CAS číslo: 71472-57-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...