2-Boc-3,4-dihydro-1h-pyrrolo[1,2-a]pyrazine-6-carboxylic acid 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR924758-250MG
OR924758-1G
APOSOR924758-250MGEA
9100
CZK
APOSOR924758-250MG
APOSOR924758-1G
2-Boc-3,4-dihydro-1h-pyrrolo[1,2-a]pyrazine-6-carboxylic acid 95%
2-Boc-3,4-dihydro-1h-pyrrolo[1,2-a]pyrazine-6-carboxylic acid
Vzorec:
C₁₃H₁₈N₂O₄ |
MDL Number:
MFCD22395729 CAS číslo: 1363380-86-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...