Tisk…

5-Amino-2-methylquinoline hydrochloride

Dodavatel: Apollo Scientific
Varování

OR72088-1G OR72088-5G
APOSOR72088-1GEA 2820 CZK
APOSOR72088-1G APOSOR72088-5G
5-Amino-2-methylquinoline hydrochloride
5-Amino-2-methylquinoline hydrochloride
Vzorec: C₁₀H₁₀N₂·ClH
CAS číslo: 2828564-69-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR