Tisk…

(1S)-(+)-2-Amino-1-phenylethan-1-ol hemihydrate

Dodavatel: Apollo Scientific
Nebezpečí

OR61461-5G OR61461-25G
APOSOR61461-5GEA 1080 CZK
APOSOR61461-5G APOSOR61461-25G
(1S)-(+)-2-Amino-1-phenylethan-1-ol hemihydrate
(1S)-(+)-2-Amino-1-phenylethan-1-ol hemihydrate
Vzorec: 2C₈H₁₁NO·H₂O
MW: 292,37 g/mol
Storage Temperature: Freezer
UN: 3259
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR