(1R)-(-)-2-Amino-1-phenylethan-1-ol hemihydrate
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR61460-5G
OR61460-25G
APOSOR61460-5GEA
1080
CZK
APOSOR61460-5G
APOSOR61460-25G
(1R)-(-)-2-Amino-1-phenylethan-1-ol hemihydrate
(1R)-(-)-2-Amino-1-phenylethan-1-ol hemihydrate
Vzorec:
2C₈H₁₁NO·H₂O Storage Temperature: Freezer |
UN:
3259 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...