2-(Difluoromethoxy)-6-fluorobenzoic acid 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC37880-250MG
PC37880-1G
APOSPC37880-250MGEA
7440
CZK
APOSPC37880-250MG
APOSPC37880-1G
2-(Difluoromethoxy)-6-fluorobenzoic acid 95%
2-(Difluoromethoxy)-6-fluorobenzoic acid
Vzorec:
C₈H₅F₃O₃ |
MDL Number:
MFCD14698505 CAS číslo: 960249-94-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...