1,19-Diamino-4,7,10,13,16-pentaoxanonadecane tech
Dodavatel: Apollo Scientific
|
Varování
|
OR55090-250MG
OR55090-1G
APOSOR55090-250MGEA
4370
CZK
APOSOR55090-250MG
APOSOR55090-1G
1,19-Diamino-4,7,10,13,16-pentaoxanonadecane tech
1,19-Diamino-4,7,10,13,16-pentaoxanonadecane
Vzorec:
C₁₄H₃₂N₂O₅ Storage Temperature: Freezer |
MDL Number:
MFCD01630929 CAS číslo: 869308-34-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...