Ethyl 7-bromo-4-oxo-4H-pyrido[1,2-a]pyrimidine-3-carboxylate
Dodavatel: Apollo Scientific
|
Varování
|
OR54821-250MG
OR54821-1G
APOSOR54821-250MGEA
622
CZK
APOSOR54821-250MG
APOSOR54821-1G
Ethyl 7-bromo-4-oxo-4H-pyrido[1,2-a]pyrimidine-3-carboxylate
Ethyl 7-bromo-4-oxo-4H-pyrido[1,2-a]pyrimidine-3-carboxylate
Vzorec:
C₁₁H₉BrN₂O₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD12546518 CAS číslo: 16867-56-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...