Tisk…

Methyl 3-bromo-2-chloro-5-iodobenzoate 95%

Dodavatel: Apollo Scientific
Varování

OR54712-250MG OR54712-1G OR54712-5G
APOSOR54712-250MGEA 5390 CZK
APOSOR54712-250MG APOSOR54712-1G APOSOR54712-5G
Methyl 3-bromo-2-chloro-5-iodobenzoate 95%
Methyl 3-bromo-2-chloro-5-iodobenzoate
Vzorec: C₈H₅BrClIO₂
MDL Number: MFCD26865671
CAS číslo: 1566999-42-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR