Tisk…

2-Chloro-5-methylpyridine-4-boronic acid 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: (2-Chloro-5-methylpyridin-4-yl)boronic acid

OR906824-250MG OR906824-1G OR906824-5G
APOSOR906824-250MGEA 6550 CZK
APOSOR906824-250MG APOSOR906824-1G APOSOR906824-5G
2-Chloro-5-methylpyridine-4-boronic acid 95%
2-Chloro-5-methylpyridine-4-boronic acid
Vzorec: C₆H₇BClNO₂
MW: 171.389 g/mol
Bod tání: 110…120 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16610704
CAS číslo: 2377605-91-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR