Tisk…

2-Bromo-5-chloroisonicotinonitrile 95%

Dodavatel: Apollo Scientific
Nebezpečí

OR47899-250MG OR47899-1G OR47899-5G
APOSOR47899-250MGEA 5460 CZK
APOSOR47899-250MG APOSOR47899-1G APOSOR47899-5G
2-Bromo-5-chloroisonicotinonitrile 95%
2-Bromo-5-chloroisonicotinonitrile
Vzorec: C₆H₂BrClN₂
MDL Number: MFCD18261449
CAS číslo: 1211537-53-9
UN: 3439
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR