2-(Cyclopropyl)thiophene-3-carbonitrile 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR45204-250MG
OR45204-1G
OR45204-5G
APOSOR45204-250MGEA
4960
CZK
APOSOR45204-250MG
APOSOR45204-1G
APOSOR45204-5G
2-(Cyclopropyl)thiophene-3-carbonitrile 95%
2-(Cyclopropyl)thiophene-3-carbonitrile
Vzorec:
C₈H₇NS Storage Temperature: Refrigerator |
MDL Number:
MFCD32219466 CAS číslo: 2654087-86-0 UN: 1993 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...