Tisk…

2-Bromo-5-iodo-3-methylbenzyl alcohol 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: (2-Bromo-5-iodo-3-methylphenyl)methanol

OR45195-250MG OR45195-1G OR45195-5G
APOSOR45195-250MGEA 5390 CZK
APOSOR45195-250MG APOSOR45195-1G APOSOR45195-5G
2-Bromo-5-iodo-3-methylbenzyl alcohol 95%
2-Bromo-5-iodo-3-methylbenzyl alcohol
Vzorec: C₈H₈BrIO
MW: 326.96 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32868532
CAS číslo: 2091676-26-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR