Tisk…

2-(Trifluoroacetyl)thiazole monohydrate

Dodavatel: Apollo Scientific
Varování

PC99407-250MG PC99407-1G PC99407-5G
APOSPC99407-250MGEA 5060 CZK
APOSPC99407-250MG APOSPC99407-1G APOSPC99407-5G
2-(Trifluoroacetyl)thiazole monohydrate
2-(Trifluoroacetyl)thiazole monohydrate
Vzorec: C₅H₂F₃NOS·H₂O
MW: 199.15 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD30742687
CAS číslo: 2097800-30-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR