4-Bromo-2-fluoro-3-(trifluoromethyl)benzaldehyde 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC99134-250MG
PC99134-1G
PC99134-5G
APOSPC99134-250MGEA
5460
CZK
APOSPC99134-250MG
APOSPC99134-1G
APOSPC99134-5G
4-Bromo-2-fluoro-3-(trifluoromethyl)benzaldehyde 95%
4-Bromo-2-fluoro-3-(trifluoromethyl)benzaldehyde
Vzorec:
C₈H₃BrF₄O MW: 271,01 g/mol |
MDL Number:
MFCD32878885 CAS číslo: 2091678-71-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...