2-Chloro-4-methoxyphenacyl bromide 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR400549-250MG
OR400549-1G
OR400549-5G
APOSOR400549-250MGEA
4960
CZK
APOSOR400549-250MG
APOSOR400549-1G
APOSOR400549-5G
2-Chloro-4-methoxyphenacyl bromide 95%
2-Chloro-4-methoxyphenacyl bromide
Vzorec:
C₉H₈BrClO₂ MW: 263,52 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD12139079 CAS číslo: 30095-51-3 UN: 2922 ADR: 8+6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...