3-(Difluoromethyl)-4-fluoroaniline 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC912796-100MG
PC912796-250MG
PC912796-1G
APOSPC912796-100MGEA
3060
CZK
APOSPC912796-100MG
APOSPC912796-250MG
APOSPC912796-1G
3-(Difluoromethyl)-4-fluoroaniline 95%
3-(Difluoromethyl)-4-fluoroaniline
Vzorec:
C₇H₆F₃N Storage Temperature: Freezer |
MDL Number:
MFCD11042620 CAS číslo: 445303-96-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...